Biotin-4-fluorescein ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
3006
AATB3006EA
152
EUR
AATB3006
Biotin-4-fluorescein ≥95%
Biotin-4-fluorescein
This bifunctional biotin-fluorescein conjugate demonstrates better binding and stronger fluorescence than biotin fluorescein.
Formel:
C₃₃H₃₂N₄O₈S |
CAS-Nummer:
1032732-74-3 ADR: 5,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...