Biotin-X NTA ≥90%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Biotin-X nitrilotriacetic acid, tripotassium salt
3009
AATB3009EA
152
EUR
AATB3009
Biotin-X NTA ≥90%
Biotin-X NTA
Biotin-X nitrilotriacetic acid (biotin-X NTA) is widely used to detect histidine-tagged proteins immobilized on nitrocellulose membranes.
Formel:
C₂₆H₄₀K₃N₅O₉S Molecular Weight: 715.98 g/mol |
CAS-Nummer:
856661-92-2 ADR: 5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...