N-Succinimidyl 6-Biotinamidohexanoate ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 6-Biotinamidohexanoic Acid N-Succinimidyl Ester, N-Biotinyl-6-aminohexanoic Acid Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanoate, N-[5-(N-Succinimidyloxycarbonyl)pentyl]biotinamide, Biotin-LC-NHS, Biotin-C5-NHS Ester

3010
AATB3010EA 120 EUR
AATB3010
N-Succinimidyl 6-Biotinamidohexanoate ≥90%
N-Succinimidyl 6-Biotinamidohexanoate

This biotin has a spacer arm of ~22 Å that reduces steric hindrance when binding several biotinylated molecules to one avidin complex; This reagent penetrates cell membranes because there is no charged group.

Formel: C₂₀H₃₀N₄O₆S
Molecular Weight: 454,54 g/mol
MDL: MFCD00065502
CAS-Nummer: 72040-63-2
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR