Biotin-PEG4-amine

Lieferant: AAT BIOQUEST

Synonyme: N-(14-Amino-3,6,9,12-tetraoxatetradecyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Amine-PEG4-Biotin, N-Biotinyl-3,6,9,12-tetraoxatetradecane-1,14-diamine, N-[2-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]ethyl]biotinamide

3013
AATB3013EA 133 EUR
AATB3013
Biotin-PEG4-amine
Biotin-PEG4-amine

Biotin PEG4 amine is a biotinylation reagent with a PEG4 spacer.


  • Stable for 12 months


Quality tested in-house

Formel: C₂₀H₃₈N₄O₆S
Molecular Weight: 462,606 g/mol
MDL: MFCD26142992
CAS-Nummer: 663171-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR