Biotin-PEG2-Amine ≥95%

Lieferant: AAT BIOQUEST
Achtung

3014
AATB3014EA 152 EUR
AATB3014
Biotin-PEG2-Amine ≥95%
Biotin-PEG2-Amine

This carbonyl-reactive biotin derivative contains a long arm (~20 Å) increase its avidin-binding affinity.

Formel: C₁₆H₃₀N₄O₄S
MDL: MFCD09264745
CAS-Nummer: 138529-46-1
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR