N-Biotinyl-N'-(3-maleimidopropionyl)-3,6-dioxaoctane-1,8-diamine ≥95%

Lieferant: AAT BIOQUEST
Achtung

3015
AATB3015EA 152 EUR
AATB3015
N-Biotinyl-N'-(3-maleimidopropionyl)-3,6-dioxaoctane-1,8-diamine ≥95%
N-Biotinyl-N'-(3-maleimidopropionyl)-3,6-dioxaoctane-1,8-diamine

This thiol-reactive biotin derivative contains a long arm (~20 angstrom) to increase its avidin-binding affinity.

Formel: C₂₃H₃₅N₅O₇S
MDL: MFCD09264743
CAS-Nummer: 305372-39-8
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR