Biotin-PEG2-NHS Biodetektion
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
N-Succinimidyl 9-(Biotinamido)-4,7-dioxanonanoate, 2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(5-(2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)ethoxy)ethoxy)propanoate, Biotin-PEG2-NHS ester, 9-(Biotinamido)-4,7-dioxanonanoic acid N-Succinimidyl ester
3016
AATB3016EA
152
EUR
AATB3016
Biotin-PEG2-NHS Biodetektion
Biotin-PEG2-NHS
This amine-reactive biotin derivative contains a long arm (~20 Å) to increase its avidin-binding affinity.
- Improved labeling efficiency
- High binding affinity
Formel:
C₂₁H₃₂N₄O₈S Molecular Weight: 500,57 g/mol |
MDL:
MFCD00467154 CAS-Nummer: 596820-83-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...