Biotin-PEG2-Azide ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
3019
AATB3019EA
152
EUR
AATB3019
Biotin-PEG2-Azide ≥95%
Biotin-PEG2-Azide
Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins.
Formel:
C₁₆H₂₈N₆O₄S Molecular Weight: 400,496 g/mol |
MDL:
MFCD26142991 CAS-Nummer: 945633-30-7 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...