5-dR6G succinimidyl ester
Lieferant: AAT BIOQUEST
302
AATB302EA 4390 EUR
AATB302
5-dR6G succinimidyl ester
5-dR6G succinimidyl ester
5-dR6G (5-Carboxy-4,7-dichlororhodamine 6G) is one of the four critical 4,7-dichloro-rhodamine dyes used as the BigDye terminators for DNA sequencing in combination with dR110, dTMR and dROX.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only