Biotin Alkyne
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-N-(prop-2-yn-1-yl)pentanamide
3021
AATB3021EA
152
EUR
AATB3021
Biotin Alkyne
Biotin Alkyne
Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins.
Formel:
C₁₃H₁₉N₃O₂S Molecular Weight: 281,37 g/mol |
MDL:
MFCD28385487 CAS-Nummer: 773888-45-2 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...