Biotin-PEG4-NHS Biodetektion
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Biotin PEG4 succinimidyl ester, 15-biotinlamino-4,7,10,13-dioxanonanoic acid N-hydroxysuccinimidyl ester, Biotin-PEG4-NHS ester, NHS-PEO4-Biotin
3022
AATB3022EA
152
EUR
AATB3022
Biotin-PEG4-NHS Biodetektion
Biotin-PEG4-NHS
Biotin PEG4 succinimidyl ester is an amine-reactive biotin derivative that contains a long arm to increase its avidin-binding affinity.
- Improved labeling efficiency
- High binding affinity
Formel:
C₂₅H₄₀N₄O₁₀S Molecular Weight: 588,67 g/mol |
MDL:
MFCD08457831 CAS-Nummer: 459426-22-3 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...