Biotin PEG3 amine Biodetektion
Lieferant: AAT BIOQUEST
Achtung
3024
AATB3024EA 307 EUR
AATB3024
Biotin PEG3 amine Biodetektion
Biotin PEG3 amine
Biotin PEG3 amine is a carbonyl-reactive biotin derivative that contains a long arm to increase its avidin-binding affinity.
  • Improved labeling efficiency
  • High binding affinity
CAS-Nummer: 194920-43-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f8b27e1b9f5f2e9a3db6cf2c1ff7a6a8ec8380b&vid=a54dbfffd2675ef04bcdf701d1d2847f4ec23d57&prid=web&clid=SAPDAM