Biotin-PEG2-Azide für die Molekularbiologie

Lieferant: AAT BIOQUEST
Achtung

3025
AATB3025EA 153 EUR
AATB3025
Biotin-PEG2-Azide für die Molekularbiologie
Biotin-PEG2-Azide

Biotin-PEG2-Azide *CAS 945633-30-7* is a non-cleavable, azide-activated biotinylation reagent designed for use in bioconjugation applications.


  • Useful for multiple applications

Formel: C₁₆H₂₈N₆O₄S
Molecular Weight: 400,496 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD26142991
CAS-Nummer: 945633-30-7
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR