SSL biotin NHS ester, ReadiCleave™

Lieferant: AAT BIOQUEST

3030
AATB3030EA 429 EUR
AATB3030
SSL biotin NHS ester, ReadiCleave™
SSL biotin NHS ester

ReadiCleave™ SSL biotin NHS ester is a cleavable biotinylating reagent which can efficiently introduce a biotin moiety to amine-containing biomolecules.


  • Stable for 12 months


AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.


Bitte beachten For Research Use Only

Molecular Weight: 504.64 g/mol
CAS-Nummer: 122266-55-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR