Biotin maleimide, ReadiView™ Biodetektion
Lieferant: AAT BIOQUEST
|
Achtung
|
3058
AATB3058EA
307
EUR
AATB3058
Biotin maleimide, ReadiView™ Biodetektion
Biotin maleimide
Biotin/avidin complexes are widely applied for a variety of biological detections.
- High fluorescence enhancement
- Sensitive detection
- Multiple applications
Formel:
C20H29N5O5S Molecular Weight: 451,54 g/mol |
CAS-Nummer:
116919-18-7 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...