1-({5-[(3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoyl}oxy)-2,5-pyrrolidindion, ReadiView™ Biodetektion
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Biotin-NHS, 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin N-hydroxysuccinimide ester, D-Biotinyl-OSu, NHS-Biotin, Biotin N-hydroxysuccinimide ester, N-(D-Biotinyloxy)succinimide
3059
AATB3059EA
152
EUR
AATB3059
1-({5-[(3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoyl}oxy)-2,5-pyrrolidindion, ReadiView™ Biodetektion
D-Biotin-N-succinimidyl ester
Biotin/avidin complexes are widely applied for a variety of biological detections.
- High fluorescence enhancement
- Sensitive detection
- Multiple applications
Formel:
C₁₄H₁₉N₃O₅S Molecular Weight: 341,38 g/mol |
MDL:
MFCD00078531 CAS-Nummer: 35013-72-0 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...