Biocytin ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: ε-Biotinoyl-L-lysine, H-Lys(biotinyl)-OH

3080
AATB3080EA 230 EUR
AATB3080
Biocytin ≥90%
Biocytin

Biocytin is invaluable as a rapidly transporting, sensitive neuronal tracer requiring little permeabilizing agents.

Formel: C₁₆H₂₈N₄O₄S
Molecular Weight: 372,484 g/mol
MDL: MFCD00077319
CAS-Nummer: 576-19-2
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR