Biocytin C2 maleimide Biodetektion

Lieferant: AAT BIOQUEST
Achtung

3085
AATB3085EA 307 EUR
AATB3085
Biocytin C2 maleimide Biodetektion
Biocytin C2 maleimide

Biocytin maleimide readily reacts with thiol moieties of biopolymers to form thioether conjugate that is quite stable.


  • Thiol-reactive
  • Multiple applications

Formel: C₂₂H₃₄N₆O₅S
Molecular Weight: 494,61 g/mol
CAS-Nummer: 1248344-33-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a57baa83316552a0c2cb9e28d915cf10d6d6a013&vid=00cccb49ecaf26157ec6835da719a85ed0dd541d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR