Biocytin hydrazide hydrochloride ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
3086
AATB3086EA
120
EUR
AATB3086
Biocytin hydrazide hydrochloride ≥95%
Biocytin hydrazide hydrochloride
This water-soluble biocytin hydrazide is used for the selective non-radioactive detection of glycoconjugates.
Formel:
C₁₆H₃₁ClN₆O₃S |
CAS-Nummer:
102743-85-1 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...