ReadiUse™ Universal Kinase Reaction Stopping Buffer
Lieferant: AAT BIOQUEST
31005
AATB31005EA
133
EUR
AATB31005
ReadiUse™ Universal Kinase Reaction Stopping Buffer
Puffer
Klonpuffer
ReadiUse™ Universal Kinase Reaction Stopping Buffer is an aqueous solution that effectively chelates divalent metal ions.
- Designed for ease of use
- Saves time
This sterile-filtered solution finds wide applicability in biochemistry and molecular biology research.
Bitte beachten For research use only.