mFluor™ UV 375 Biotin Konjugat
Lieferant: AAT BIOQUEST
3111
AATB3111EA 309 EUR
AATB3111
mFluor™ UV 375 Biotin Konjugat
mFluor™ UV 375 Biotin
mFluor™ UV 375 Biotin Conjugate contains mFluor™ UV 375 fluorophore that is well excited by the common 350 nm laser.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only