mFluor™ UV 460 Biotin Konjugat
Lieferant: AAT BIOQUEST
3112
AATB3112EA 309 EUR
AATB3112
mFluor™ UV 460 Biotin Konjugat
mFluor™ UV 460 Biotin
mFluor™ UV 460 Biotin Conjugate contains mFluor™ UV 460 fluorophore that is well excited by the common 350 nm laser.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only