mFluor™ Violet 500 Biotin Konjugat
Lieferant: AAT BIOQUEST
3113
AATB3113EA 309 EUR
AATB3113
mFluor™ Violet 500 Biotin Konjugat
mFluor™ Violet 500 Biotin
mFluor™ Violet 500 Biotin Conjugate contains mFluor™ Violet 500 fluorophore that is well excited by the common 405 nm violet laser.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only