mFluor™ Violet 540 Biotin Konjugat
Lieferant: AAT BIOQUEST
3114
AATB3114EA 309 EUR
AATB3114
mFluor™ Violet 540 Biotin Konjugat
mFluor™ Violet 540 Biotin
mFluor™ Violet 540 Biotin Conjugate contains mFluor™ Violet 540 fluorophore that is well excited by the common 405 nm violet laser.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only