mFluor™ Red 780 Biotin Konjugat
Lieferant: AAT BIOQUEST
mFluor™ Red 780 Biotin Conjugate contains mFluor™ Red 780 fluorophore that is well excited by the common 350 nm laser.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only