mFluor™ Violet 450-PEG4-Biotin Konjugat
Lieferant: AAT BIOQUEST
3116
AATB3116EA 271 EUR
AATB3116
mFluor™ Violet 450-PEG4-Biotin Konjugat
mFluor™ Violet 450-PEG4-Biotin
mFluor™ Violet 450-PEG4-Biotin Conjugate contains mFluor™ Violet 450 fluorophore that is well excited by the common 405 nm violet laser.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only