5-dR110 succinimidyl ester ≥75%
Lieferant: AAT BIOQUEST
317
AATB317EA
7310
EUR
AATB317
5-dR110 succinimidyl ester ≥75%
5-dR110 succinimidyl ester
Compared to fluorescein labeling reagents such as FITC and FAM, 5-dR110 [5-carboxy-4,7-dichlororhodamine 110] reagents give more photostable and pH-independent bioconjugates that have the absorption maximum around the preferred 488 nm excitation wavelength.
- Sanger sequencing building block
Bitte beachten For research use only.