5-Carboxyrhodamine 110 (5-CR110) einfaches Isomer ≥95%
Lieferant: AAT BIOQUEST
322
AATB322EA
460
EUR
AATB322
5-Carboxyrhodamine 110 (5-CR110) einfaches Isomer ≥95%
5-Carboxyrhodamine 110 (5-CR110)
Compared to fluorescein labeling reagents such as FITC and FAM, CR110 reagents give more photostable and pH-independent bioconjugates that have the absorption maximum around the preferred 488 nm excitation wavelength.
- Photostable
- pH-independent Fluorescence
Bitte beachten For research use only.