6-Carboxyrhodamine 110 (6-CR110) einfaches Isomer ≥95%
Lieferant: AAT BIOQUEST
Synonyme:
Rhodamine Green™ Carboxylic Acid
323
AATB323EA
460
EUR
AATB323
6-Carboxyrhodamine 110 (6-CR110) einfaches Isomer ≥95%
6-Carboxyrhodamine 110 (6-CR110)
Compared to fluorescein labeling reagents such as FITC and FAM, CR110 reagents give more photostable and pH-independent bioconjugates that have the absorption maximum around the preferred 488 nm excitation wavelength.
- Photostable
- pH-independent Fluorescence
Bitte beachten For research use only.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...