5-(6)-Carboxyrhodamine 6G [5(6)-CR6G]
Lieferant: AAT BIOQUEST
330
AATB330EA 307 EUR
AATB330
5-(6)-Carboxyrhodamine 6G [5(6)-CR6G]
5-(6)-Carboxyrhodamine 6G [5(6)-CR6G]
In general, rhodamine 6G conjugates have higher fluorescence quantum yields than tetramethylrhodamine conjugates, as well as good photostability.
  • Bright Fluorescence
  • High Photostability
  • Useful For Small Molecule Labeling
Bitte beachten For research use only.