Reversible Biotin maleimide, ReadiLeave™
Lieferant: AAT BIOQUEST
3402
AATB3402EA 266 EUR
AATB3402
Reversible Biotin maleimide, ReadiLeave™
Reversible Biotin maleimide
ReadiLeave™ Reversible (RLR) Biotin is a newly developed biotin derivative that has significantly reduced affinity to avidin (including streptavidin) to make the binding of RLR biotin and streptavidin readily reversible when needed.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only