5(6)-Carboxyrhodamine 6G cadaverine ≥95%
Lieferant: AAT BIOQUEST
343
AATB343EA
460
EUR
AATB343
5(6)-Carboxyrhodamine 6G cadaverine ≥95%
5(6)-Carboxyrhodamine 6G cadaverine
5(6)-Caroxyrhodamine 6G cadaverine is a building block for developing rhodamine bioconjugates.
- Useful for Multicolor Fluorescence Imaging
- High Photostability
- Improved Fluorescence Quantum Yield
Bitte beachten For research use only.