5(6)-Carboxyrhodamine 6G ethylenediamine ≥95%
Lieferant: AAT BIOQUEST
344
AATB344EA
460
EUR
AATB344
5(6)-Carboxyrhodamine 6G ethylenediamine ≥95%
5(6)-Carboxyrhodamine 6G ethylenediamine
5(6)-Carboxyrhodamine 6G ethylenediamine is a building block for developing rhodamine bioconjugates.
- Useful for Multicolor Fluorescence Imaging
- High Photostability
- Improved Fluorescence Quantum Yield
Bitte beachten For research use only.