5-CR6G, SE einfaches Isomer ≥90%

Lieferant: AAT BIOQUEST

Synonyme: 5-CR6G, NHS ester, 5-Carboxyrhodamine 6G, succinimidyl ester

345
AATB345EA 307 EUR
AATB345
5-CR6G, SE einfaches Isomer ≥90%
5-CR6G, SE

5-CR6G, SE is amine-reactive derivative of 5-CR6G.


  • Useful for Multicolor Fluorescence Imaging
  • High Photostability
  • Improved Fluorescence Quantum Yield


Bitte beachten For research use only.

Formel: C₃₁H₂₉N₃O₇
Molecular Weight: 555,59 g/mol
CAS-Nummer: 209112-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR