Digoxigenin NHS ester Biodetektion

Lieferant: AAT BIOQUEST
Achtung

3500
AATB3500EA 230 EUR
AATB3500
Digoxigenin NHS ester Biodetektion
Digoxigenin NHS ester

Digoxigenin (DIG) is a hapten, a small molecule with high antigenicity, which is used in many biological detections similarly to other popular haptens such as 2,4-Dinitrophenol (DNP) and biotin.


  • Improved labeling efficiency
  • High binding affinity

Formel: C₃₅H₅₀N₂O₁₀
Molecular Weight: 658,79 g/mol
CAS-Nummer: 129273-26-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=018eb772b45cfb777a506932e46934e166e7fd69&vid=3d80d1cb17bbb322925a58b7b5109722c17ecbae&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR