Digoxigenin NHS ester

Lieferant: AAT BIOQUEST

3501
AATB3501EA 918 EUR
AATB3501
Digoxigenin NHS ester
Digoxigenin NHS ester

Digoxigenin (DIG) is a hapten, a small molecule with high antigenicity, which is used in many biological detections similarly to other popular haptens such as 2,4-Dinitrophenol (DNP) and biotin.


  • Stable for 12 months


Quality tested in-house


Bitte beachten For Research Use Only

Formel: C₃₅H₅₀N₂O₁₀
CAS-Nummer: 129273-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR