Digoxigenin maleimide Biodetektion
Lieferant: AAT Bioquest
Achtung
3503
AATB3503EA 307 EUR
AATB3503
Digoxigenin maleimide Biodetektion
Digoxigenin maleimide
Digoxigenin (DIG) is a hapten, a small molecule with high antigenicity, which is used in many biological detections similarly to other popular haptens such as 2,4-Dinitrophenol (DNP) and biotin.
  • Thiol-reactive
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c300f50ac87b799065bdc584c38a35f7fcbd03ce&vid=ad2621b34048a6aa97890ca950f04625cbf1b9b1&prid=web&clid=SAPDAM