Digoxigenin azide Biodetektion
Lieferant: AAT BIOQUEST
Achtung
3504
AATB3504EA 307 EUR
AATB3504
Digoxigenin azide Biodetektion
Digoxigenin azide
Digoxigenin (DIG) is a hapten, a small molecule with high antigenicity, which is used in many biological detections similarly to other popular haptens such as 2,4-Dinitrophenol (DNP) and biotin.
  • Improved labeling efficiency
  • High binding affinity
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0289f5589732ee0dacaf1e0a317cc2bc52362476&vid=6b16a65e4cd3473055f468cbcd66b5fcf5167dfe&prid=web&clid=SAPDAM