Digoxigenin azide Biodetektion
Lieferant: AAT BIOQUEST
|
Achtung
|
3504
AATB3504EA
307
EUR
AATB3504
Digoxigenin azide Biodetektion
Digoxigenin azide
Digoxigenin (DIG) is a hapten, a small molecule with high antigenicity, which is used in many biological detections similarly to other popular haptens such as 2,4-Dinitrophenol (DNP) and biotin.
- Improved labeling efficiency
- High binding affinity
ADR:
5,II |