Digoxigenin alkyne Biodetektion
Lieferant: AAT BIOQUEST
Achtung
3505
AATB3505EA 307 EUR
AATB3505
Digoxigenin alkyne Biodetektion
Digoxigenin alkyne
Digoxigenin (DIG) is a hapten, a small molecule with high antigenicity, which is used in many biological detections similarly to other popular haptens such as 2,4-Dinitrophenol (DNP) and biotin.
  • Improved labeling efficiency
  • High binding affinity
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be10d43fe5f4dc315823e51d43906a25da5344d1&vid=96413c20c17a21b4b34cf979434370bd6a5b7cca&prid=web&clid=SAPDAM