Digoxigenin C9 acid
Lieferant: AAT BIOQUEST
3510
AATB3510EA
420
EUR
AATB3510
Digoxigenin C9 acid
Digoxigenin C9 acid
Digoxigenin (DIG) is a hapten, a small molecule with high antigenicity, which is used in many biological detections similarly to other popular haptens such as 2,4-Dinitrophenol (DNP) and biotin.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only