5(6)-TAMRA cadaverine ≥95%
Lieferant: AAT BIOQUEST
Synonyme: Tetramethylrhodamine 5-(and-6)-carboxamide cadaverine
355
AATB355EA 460 EUR
AATB355
5(6)-TAMRA cadaverine ≥95%
5(6)-TAMRA cadaverine
5(6)-TAMRA cadaverine is a building block for developing fluoresceinated bioconjugates.
  • Multiple Bioconjugation Applications
  • 95% Purity
Bitte beachten For research use only.