5-TAMRA cadaverine ≥95%
Lieferant: AAT BIOQUEST
Synonyme:
Tetramethylrhodamine-5-carboxamide cadaverine
356
AATB356EA
460
EUR
AATB356
5-TAMRA cadaverine ≥95%
5-TAMRA cadaverine
5-TAMRA cadaverine is a building block for developing fluoresceinated bioconjugates.
- Multiple Bioconjugation Applications
- 95% Purity
Bitte beachten For research use only.