Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit *Optimized for microplate reader*
Lieferant: AAT BIOQUEST
36312
AATB36312EA 614 EUR
AATB36312
Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit *Optimized for microplate reader*
Tests Zelltests
Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit enables homogeneous fluorescence-based assays for detecting intracellular calcium mobilization without the need to use kinetics reading mode.
  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Optimized for microplate readers but suitable for all major fluorescence platforms
  • Multiple research applications
Bitte beachten For research use only.