Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit *Optimized for microplate reader*
Lieferant: AAT BIOQUEST
36312
AATB36312EA
614
EUR
AATB36312
Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit *Optimized for microplate reader*
Tests
Zelltests
Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit enables homogeneous fluorescence-based assays for detecting intracellular calcium mobilization without the need to use kinetics reading mode.
- Ready to use
- Contains all necessary components
- Robust and adaptable
- Optimized for microplate readers but suitable for all major fluorescence platforms
- Multiple research applications
Bitte beachten For research use only.