5-Carboxytetramethylrhodamine (5-TAMRA) fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 5-TAMRA, 5-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate, 2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxybenzoate, TAMRA, 5-isomer

365
AATB365EA 3690 EUR
AATB365
5-Carboxytetramethylrhodamine (5-TAMRA) fluoreszierende Probe
5-Carboxytetramethylrhodamine (5-TAMRA)

5-TAMRA is the purified single isomer of 5(6)-TAMRA. It is widely used to label peptides and proteins. It is preferred for some complicated biological applications where reproducibility is more critical than material cost since the minor positional difference between 5-TAMRA and 6-TAMRA might affect some biological properties of the underlying conjugates. Currently AAT Bioquest is using a proprietary process to synthesize isomer-free 5-TAMRA without the conventional chromatographic purification.


  • High reproducibility

Formel: C₂₅H₂₂N₂O₅
Molecular Weight: 430,45 g/mol
MDL: MFCD00269769
CAS-Nummer: 91809-66-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a69abfcb05257a3d0e6e5d00246543a8c27ab383&vid=b096118283475b5bdd0437820922f5d2a524ed10&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR