CLF [Cholyl-Lys-Fluorescein]

Lieferant: AAT Bioquest
Achtung

36701
AATB36701EA 460 EUR
AATB36701
CLF [Cholyl-Lys-Fluorescein]
CLF [Cholyl-Lys-Fluorescein]

CLF is a fluorescein-labeled bile acid analog that closely parallels the cellular binding and uptake properties of cholic acid derivatives.


  • Well-cited
  • Stable signal
  • Multiple applications

Lagertemperatur: Gefrierschrank
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR