2-NBDG ≥90%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, NBD-Glucose, (2R,3R,4S,5R)-3,4,5,6-Tetrahydroxy-2-((7-nitrobenzo[c][1,2,5]oxadiazol-4-yl)amino)hexanal
36702
AATB36702EA
230
EUR
AATB36702
2-NBDG ≥90%
2-NBDG
2-NBDG is a fluorescently-labeled deoxyglucose analog that is used primarily to directly monitor glucose uptake by living cells and tissues.
Formel:
C₁₂H₁₄N₄O₈ Molecular Weight: 342,3 g/mol |
MDL:
MFCD23135882 CAS-Nummer: 186689-07-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...