Cholyl-iFluor™ 647 Konjugat

Lieferant: AAT BIOQUEST
Achtung

36703
AATB36703EA 439 EUR
AATB36703
Cholyl-iFluor™ 647 Konjugat
Cholyl-iFluor™ 647

Cholyl-iFluor® 647 conjugate is a red fluorescent analog of our green fluorescent Cholyl-Lys-Fluorescein (#36701).


  • Bright signal
  • Long-wavelength emission
  • Suitable for multiplexing

Molecular Weight: 1289,57 g/mol
Lagertemperatur: Gefrierschrank
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR