6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 6-Carboxytetramethylrhodamine N-succinimidyl ester, 6-TAMRA, SE, 6-TAMRA N-succinimidyl ester

376 377 378
AATB376EA 159 EUR
AATB376 AATB377 AATB378
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) ≥90%
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE)

The single TAMRA isomers are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purification that is often required in the conjugation processes.

Formel: C₂₉H₂₅N₃O₇
Molecular Weight: 527.52 g/mol
CAS-Nummer: 150810-69-8
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58266059a983bf709d98cefa90c90f15d1a35310&vid=87fef656765193135897bbee96ef4d73f827ba19&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR