5(6)-Carboxy-X-rhodamine ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 9-[2,4(or 2,5)-Dicarboxyphenyl]-2,3,6,7,12,13,16,17-octahydro-1H,5H,11H,15H-xantheno[2,3,4-ij:5,6,7-i'j′]diquinolizin-18-ium inner salt, 5(6)-ROX

380
AATB380EA 769 EUR
AATB380
5(6)-Carboxy-X-rhodamine ≥95%
5(6)-Carboxy-X-rhodamine

ROX dyes are strongly red fluorescent.

Formel: C₃₃H₃₀N₂O₅
Molecular Weight: 534,612 g/mol
MDL: MFCD01862979
CAS-Nummer: 198978-94-8
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR