6-ROX ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 6-Carboxy-X-rhodamine, Carboxy-X-rhodamine, 6-isomer

382
AATB382EA 230 EUR
AATB382
6-ROX ≥95%
6-ROX

6-ROX is the other purified single isomer of 5(6)-ROX.

Formel: C₃₃H₃₀N₂O₅
Molecular Weight: 534.6 g/mol
CAS-Nummer: 194785-18-7
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR